Learn More
Cayman Chemical 1-Azakenpaulon 10mg
SDP
Catalog No. 502966333
Encompass_Preferred
Inhibits GSK3β more potently (IC50 = 18 nM) than CDK1/cyclin B or CDK5/p25 (IC50s = 2.0 and 4.2 µM, respectively); stimulates the proliferation of human islets and drives the differentiation of mouse embryonic stem cells and Nematostella larvae
Catalog No. 50-296-6333 Supplier Cayman Chemical Supplier No. 1673310 MG
Inhibits GSK3β more potently (IC50 = 18 nM) than CDK1/cyclin B or CDK5/p25 (IC50s = 2.0 and 4.2 µM, respectively); stimulates the proliferation of human islets and drives the differentiation of mouse embryonic stem cells and Nematostella larvae