Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Azaphenothiazine 98.0+%, TCI America™
SDP

Catalog No. A28621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28621G 1 g
A2862200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28621G Supplier TCI America Supplier No. A28621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 261-96-1
Molecular Formula C11H8N2S
Molecular Weight (g/mol) 200.259
MDL Number MFCD00160449
InChI Key UKDZROJJLPDLDO-UHFFFAOYSA-N
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine
PubChem CID 67496
IUPAC Name 10H-pyrido[3,2-b][1,4]benzothiazine
SMILES C1=CC=C2C(=C1)NC3=C(S2)C=CC=N3

Specifications

Melting Point 112°C
Color Yellow
Boiling Point 370°C
Quantity 1 g
Formula Weight 200.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Azaphenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.