Learn More
Medchemexpress LLC 1-azido-20-bromo-3,6,9,12,15,18-hexaoxaicosane | 2062663-64-1 | 95.0% | C14H28BrN3O6 | 10MG
SDP
Catalog No. 5000258802
Encompass_Preferred
Bromo-PEG6-azide is a PEG-based bifunctional linker bearing a terminal azide and a bromide. It is intended for use in PROTAC assembly and bioconjugation workflows, enabling click chemistry and nucleophilic substitution while improving solubility through a six-unit PEG spacer.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Bromide provides a handle for nucleophilic substitution or alkylation reactions.
  • PEG6 spacer increases aqueous solubility and reduces aggregation.
  • Flexible spacer length aids in linker design and biological presentation.
  • High stated purity suitable for synthetic applications.
  • Defined molecular weight simplifies analytical characterization.
Catalog No. 50-002-58802 Supplier Medchemexpress LLC Supplier No. HY14082310MG
Bromo-PEG6-azide is a PEG-based bifunctional linker bearing a terminal azide and a bromide. It is intended for use in PROTAC assembly and bioconjugation workflows, enabling click chemistry and nucleophilic substitution while improving solubility through a six-unit PEG spacer.
  • Terminal azide enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Bromide provides a handle for nucleophilic substitution or alkylation reactions.
  • PEG6 spacer increases aqueous solubility and reduces aggregation.
  • Flexible spacer length aids in linker design and biological presentation.
  • High stated purity suitable for synthetic applications.
  • Defined molecular weight simplifies analytical characterization.