Learn More
Medchemexpress LLC 1-azido-3,6,9,12,15,18,21,24-octaoxaheptacosane-27-hydrazide | 2353410-11-2 | MFCD31693701 | 96.1% | C19H39N5O9 | 10MG
SDP
Catalog No. 5000261536
Encompass_Preferred
Azido-PEG8-hydrazide is a heterobifunctional PEG-based linker bearing an azide group and a terminal hydrazide. Supplied as a solid reagent for research use, it enables selective bioconjugation via click chemistry or hydrazone formation and is suitable for assembling linkers in small-molecule conjugates and PROTACs.
  • Contains an azide group for copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Features a hydrazide terminus for hydrazone formation with carbonyl partners.
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance.
  • Supplied as a stable solid suitable for small-scale synthesis and handling.
  • High purity supports reproducible conjugation reactions.
  • Compatible with common organic solvents; store sealed and protect from moisture for best stability.
Catalog No. 50-002-61536 Supplier Medchemexpress LLC Supplier No. HY14081510MG
Azido-PEG8-hydrazide is a heterobifunctional PEG-based linker bearing an azide group and a terminal hydrazide. Supplied as a solid reagent for research use, it enables selective bioconjugation via click chemistry or hydrazone formation and is suitable for assembling linkers in small-molecule conjugates and PROTACs.
  • Contains an azide group for copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Features a hydrazide terminus for hydrazone formation with carbonyl partners.
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance.
  • Supplied as a stable solid suitable for small-scale synthesis and handling.
  • High purity supports reproducible conjugation reactions.
  • Compatible with common organic solvents; store sealed and protect from moisture for best stability.