Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Azido-4-Bromobenzene Solution

Catalog No. 77937710ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-937-710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 77-937-710ML Supplier Sigma Aldrich Supplier No. 77937710ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Azido-4-Bromobenzene Solution

Specifications

CAS 384-67-8
Density 1.293 g/mL at 25°C, 1.3178 g/mL at 25°C
Boiling Point 94°C to 96°C35 mmHg, lit.)
Molecular Formula C8H5ClF2O
Linear Formula C6H5COCF2Cl
Quantity 10 mL
MDL Number MFCD00042123
Molecular Weight (g/mol) 190.57 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.