Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aziridineethanol 97%, Thermo Scientific™

Catalog No. AC148021000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148021000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC148021000 Supplier Thermo Scientific Chemicals Supplier No. 148021000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-52-2
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
InChI Key VYONOYYDEFODAJ-UHFFFAOYSA-N
Synonym 1-aziridineethanol, 2-aziridin-1-yl ethanol, 2-1-aziridinyl ethanol, 1-azirdineethanol, 2-azidin-1-yl ethanol, 1-2-hydroxyethyl ethylenimine, n-2-hydroxyethyl aziridine, ethanol, 2-1-aziridinyl, n-2-hydroxyethyl ethylenimine, n-2-hydroxyethyl ethyleneimine
PubChem CID 14074
ChEBI CHEBI:82393
IUPAC Name 2-(aziridin-1-yl)ethanol
SMILES C1CN1CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Quantity 100 g
Formula Weight 87.12
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1-Aziridineethanol, 97%
Hazard Category CANCER SUSPECTED AGENT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.