Learn More
Apexbio Technology LLC 1-B-D-ARAF URACIL-200mg
SDP
Catalog No. 5000897472
Encompass_Preferred
1-beta-D-Arabinofuranosyluracil (CAS No 3083-77-0) is a compound frequently studied in the field of antiviral and anticancer research Originating from modification of natural nucleosides it incorporates into nucleic acids thereby disrupting DNA synthesis and function Its mechanism of action involves the inhibition of viral replication by targeting viral polymerases and influencing cellular pathways that regulate DNA metabolism In vitro studies have demonstrated its activity in the nanomolar to low micromolar range showing potent inhibition of viral replication and DNA synthesis in various cancer cell lines It is commonly used in cell models to evaluate antiviral efficacy and in animal studies for assessing therapeutic potential This compound is also employed in drug screening assays to identify possible inhibitors of nucleic acid synthesis Experimental concentrations typically depend on the specific research design allowing for flexibility in various study protocols
Catalog No. 50-008-97472 Supplier Apexbio Technology LLC Supplier No. C6637200
1-beta-D-Arabinofuranosyluracil (CAS No 3083-77-0) is a compound frequently studied in the field of antiviral and anticancer research Originating from modification of natural nucleosides it incorporates into nucleic acids thereby disrupting DNA synthesis and function Its mechanism of action involves the inhibition of viral replication by targeting viral polymerases and influencing cellular pathways that regulate DNA metabolism In vitro studies have demonstrated its activity in the nanomolar to low micromolar range showing potent inhibition of viral replication and DNA synthesis in various cancer cell lines It is commonly used in cell models to evaluate antiviral efficacy and in animal studies for assessing therapeutic potential This compound is also employed in drug screening assays to identify possible inhibitors of nucleic acid synthesis Experimental concentrations typically depend on the specific research design allowing for flexibility in various study protocols