Learn More
Cml Supply 1 BAG OF 1000 23G 1/2 INCH BL
Catalog No. NC9057380
Encompass_Preferred
Tip, Needle; Blunt; Disposable; 304SS Tubing and PP luer- lock hub; Color Coded for Easy Recognition; 23g x 1/2 in.; 1000/BG
Catalog No. NC9057380 Supplier Cml Supply Supplier No. 90123050M
Tip, Needle; Blunt; Disposable; 304SS Tubing and PP luer- lock hub; Color Coded for Easy Recognition; 23g x 1/2 in.; 1000/BG