Learn More
Chemglass Life Sciences 1" Beaded Pipe to 1/2" Female NPT Aluminum Adapter
SDP
Catalog No. CG196440
Encompass_Preferred
1" Beaded Pipe to 1/2" Female NPT Aluminum Adapter. Aluminum transition adapter from beaded pipe on the inlet/outlet of the reactor to NPT. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.
Catalog No. CG196440 Supplier Chemglass Life Sciences Supplier No. CG196440
1" Beaded Pipe to 1/2" Female NPT Aluminum Adapter. Aluminum transition adapter from beaded pipe on the inlet/outlet of the reactor to NPT. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.