Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzhydryl-3-azetidinone 96.0+%, TCI America™
SDP

Catalog No. B47995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47995G 5 g
B47991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47995G Supplier TCI America Supplier No. B47995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40320-60-3
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00964539
InChI Key AVUDXLOVIBJFQA-UHFFFAOYSA-N
Synonym 1-diphenylmethyl azetidin-3-one, 1-benzhydryl-azetidin-3-one, 1-1,1-diphenylmethyl azetidin-3-one, 1-diphenylmethyl-3-azetidinone, 1-benzhydryl-3-azetidinone, 3-azetidinone, 1-diphenylmethyl, pubchem10151, acmc-209jd2, n-benzhydrylazetidin-3-one
PubChem CID 3735558
IUPAC Name 1-benzhydrylazetidin-3-one
SMILES C1C(=O)CN1C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 355°C
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzhydryl-3-azetidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.