Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzhydryl-3-azetidinyl Methanesulfonate 95.0+%, TCI America™
SDP

Catalog No. D41311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41311G Supplier TCI America Supplier No. D41311G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33301-41-6
Molecular Formula C17H19NO3S
Molecular Weight (g/mol) 317.40
MDL Number MFCD00159216
InChI Key MSVZMUILYMLJCF-UHFFFAOYSA-N
Synonym 1-benzhydryl-3-methanesulfonyloxy azetidine, 1-benzhydryl-3-methanesulfonatoazetidine, 1-diphenylmethyl-3-methanesulfonyloxy azetidine, 1-diphenylmethyl-3-azetidinyl methanesulfonate, 1-diphenylmethyl azetidin-3-yl methanesulfonate, 1-benzhydryl-3-azetidinyl methanesulfonate, 1-benzhydrylazetidin-3-yl methanesulfonate, methanesulfonic acid 1-benzhydryl-azetidin-3-yl ester, 1-benzhydryl-3-methanesulfonyloxyazetidine
PubChem CID 2758716
IUPAC Name 1-(diphenylmethyl)azetidin-3-yl methanesulfonate
SMILES CS(=O)(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 111°C
Color White
Quantity 1 g
Formula Weight 317.40
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzhydryl-3-azetidinyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.