Learn More
Medchemexpress LLC 1-Benzhydryl-4-cinnamylpiperazine-2,2,3,3,5,5,6,6-d8 | 1185242-27-6 | 98.2% | 5 MG
SDP
Catalog No. 5000414181
Encompass_Preferred
Cinnarizine-d8 is a deuterium labeled Cinnarizine, functioning as an antihistamine and calcium channel blocker.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-14181 Supplier Medchemexpress LLC Supplier No. HYB1090S5MG
Cinnarizine-d8 is a deuterium labeled Cinnarizine, functioning as an antihistamine and calcium channel blocker.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS