Learn More
Medchemexpress LLC 1-benzopyrylium, 7-hydroxy-5-methoxy-6-methyl-2-phenyl-, perchlorate (1:1) | 125536-25-6 | 366.75 | 1 MG
SDP
Catalog No. 5000417096
Encompass_Preferred
Dracorhodin perchlorate is a natural product derived from Dragon's blood. It acts as a pro-apoptotic compound, regulating TLR4, promoting wound healing, and exhibiting anti-tumor activity against various cancers.
  • Acts as a pro-apoptotic compound
  • Inhibits PI3K/Akt and NF-κB activation
  • Upregulates p53 expression and activates caspase
  • Produces reactive oxygen species (ROS)
  • Regulates TLR4 pathway
  • Promotes wound healing and improves diabetes
  • Exhibits anti-tumor activity against various cancers, including prostate, breast, and cervical cancer
Catalog No. 50-004-17096 Supplier Medchemexpress LLC Supplier No. HYN07261MG
Dracorhodin perchlorate is a natural product derived from Dragon's blood. It acts as a pro-apoptotic compound, regulating TLR4, promoting wound healing, and exhibiting anti-tumor activity against various cancers.
  • Acts as a pro-apoptotic compound
  • Inhibits PI3K/Akt and NF-κB activation
  • Upregulates p53 expression and activates caspase
  • Produces reactive oxygen species (ROS)
  • Regulates TLR4 pathway
  • Promotes wound healing and improves diabetes
  • Exhibits anti-tumor activity against various cancers, including prostate, breast, and cervical cancer