Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophen-5-yl isocyanate, 97%, Thermo Scientific™

Catalog No. CC24806DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC24806DE 5 g
CC24806CB 250 mg
CC24806DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC24806DE Supplier Thermo Scientific Chemicals Supplier No. CC24806DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 239097-78-0
Molecular Formula C9H5NOS
Molecular Weight (g/mol) 175.205
InChI Key GWLZJAWTWVSHCU-UHFFFAOYSA-N
Synonym 1-benzothiophen-5-yl isocyanate, 1-benzothiophen-5-ylisocyanate, benzo b thiophen-5-isocyanate, 5-isocyanato-benzo b thiophene, benzo b thiophene,5-isocyanato
PubChem CID 7060535
IUPAC Name 5-isocyanato-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 175.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophen-5-yl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.