Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC12204DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12204DA 1 g Amber Glass Bottle
CC12204EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12204DA Supplier Thermo Scientific Chemicals Supplier No. CC12204DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-carbaldehyde
Hazard Category Warning
Hazard Statement GHS P Statement Causes serious eye irritation.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.