Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophene-3-carboximidamidine hydrochloride hydrate, 97%, Thermo Scientific™

Catalog No. CC12331EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12331EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12331EA Supplier Thermo Scientific Chemicals Supplier No. CC12331EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 465515-36-0
Molecular Formula C9H11ClN2OS
Molecular Weight (g/mol) 230.71
MDL Number MFCD03659730
InChI Key DCDNRDYUSJNDGA-UHFFFAOYSA-N
Synonym 1-benzothiophene-3-carboximidamide hydrochloride, 1-benzothiophene-3-carboximidamidine hydrochloride, benzo b thiophene-3-carboxamidine, chloride
PubChem CID 2776350
IUPAC Name 1-benzothiophene-3-carboximidamide;hydrate;hydrochloride
SMILES O.Cl.NC(=N)C1=CSC2=CC=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 212.7
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carboximidamidine hydrochloride
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.