Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-benzothiophene-3-sulfonyl chloride

Catalog No. CDS014505 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014505 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014505 Supplier Sigma Aldrich Supplier No. CDS014505100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylformamide diethyl acetal undergoes me3sicl-mediated three-component coupling reaction with alkyne to yield 2,3,4,5-tetrasubstituted pyridine derivatives.n,n-dimethylformamide diethyl acetal is a suitable reagent for the esterification of fatty acids
  • It is suitable reagent used in the quantification of cocaine and its primary metabolite, benzoyl ecgonine, from a urine matrix by gas chromatography.reagent for the esterification of fatty acids; review

Specifications

Molecular Formula C8H5ClO2S2
Linear Formula C8H5ClO2S2
MDL Number MFCD02681897
Quantity 100 mL
Molecular Weight (g/mol) 232.71
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.