Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-1-phenylhydrazine 98.0+%, TCI America™
SDP

Catalog No. B115710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B115710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B115710G Supplier TCI America Supplier No. B115710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 614-31-3
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD00059186
InChI Key SQMOOVFBFVTTGF-UHFFFAOYSA-N
PubChem CID 69192
IUPAC Name 1-benzyl-1-phenylhydrazine
SMILES C1=CC=C(C=C1)CN(C2=CC=CC=C2)N

Specifications

Boiling Point 218°C
Quantity 10 g
Formula Weight 198.27
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-1-phenylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.