Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Benzyl 2-methyl (2S,4R)-4-((4-bromoisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate

Catalog No. 901735100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-173-5100MG 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-173-5100MG Supplier Sigma Aldrich Supplier No. 901735100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Terpinolene is a biogenically emitted monoterpene
  • Kinetics of gas phase reaction of oh and no3 radicals with terpinolene has been investigated
  • Biotransformation of terpinolene byaspergillus niger and several related species has been reported
  • Epoxidation of terpinolene in anhydrous conditions using alumina as a heterogeneous catalyst and hydrogen peroxide as the oxidant agent has been studied
  • It is an major component of essential oil extracted from the aerial parts of clausena indica

Specifications

CAS 1269195-05-2
Melting Point 99°C
Molecular Formula C23H23BrN2O6
Linear Formula C23H23BrN2O6
Quantity 500 mL
Synonym Aryl halide chemistry informer library compound X8
Molecular Weight (g/mol) 503.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.