Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonylchloride hydrochloride, ≥97%, Thermo Scientific™

Catalog No. CC28902DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC28902DE 5 g
CC28902DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC28902DE Supplier Thermo Scientific Chemicals Supplier No. CC28902DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 568577-86-6
Molecular Formula C12H12Cl2N2OS
Molecular Weight (g/mol) 303.201
MDL Number MFCD04974045
InChI Key AUZZRVMDIAHPNZ-UHFFFAOYSA-N
Synonym 1-benzyl-2-methylsulfanyl-1h-imidazole-5-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanyl imidazole-4-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride hydrochloride, 1-benzyl-2-methylthio-1h-imidazole-5-carbonyl chloride hydrochloride
PubChem CID 2795486
IUPAC Name 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride;hydrochloride
SMILES CSC1=NC=C(N1CC2=CC=CC=C2)C(=O)Cl.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 303.21
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonyl chloride hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.