Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%

Catalog No. AC105901000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105901000 100 g Glass bottle
AC105900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105901000 Supplier Thermo Scientific Chemicals Supplier No. 105901000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1454-53-1 | C15H20ClNO3 | 297.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-53-1
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012800
InChI Key YPFMNHZRNXPYBG-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride
PubChem CID 102623
IUPAC Name ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160.0°C to 170.0°C
Color Cream to Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 100 g
Beilstein 22, II, 216
Formula Weight 297.77
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-929-9
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.