Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-hydroxypiperidine 97.0+%, TCI America™
SDP

Catalog No. B48685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48685G 5 g
B486825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48685G Supplier TCI America Supplier No. B48685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14813-01-5
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00023734
InChI Key UTTCOAGPVHRUFO-UHFFFAOYNA-N
Synonym 1-benzyl-3-piperidinol, 1-benzyl-3-hydroxypiperidine, n-benzyl-3-hydroxypiperidine, 1-n-benzyl-3-hydroxy-piperidine, 3-piperidinol, 1-phenylmethyl, n-benzyl-3-piperidinol, pubchem7640, pubchem7727, acmc-209xh5, acmc-209xj8
PubChem CID 85773
IUPAC Name 1-benzylpiperidin-3-ol
SMILES OC1CCCN(CC2=CC=CC=C2)C1

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 g
Formula Weight 191.27
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-hydroxypiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.