Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-piperidone hydrochloride hydrate

Catalog No. AAA1284914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1284914 25 g
AAA1284906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1284914 Supplier Thermo Scientific Chemicals Supplier No. A1284914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 50606-58-1 | C12H16ClNO | 225.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50606-58-1
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD00150096
InChI Key OVWSFXNSJDMRPV-UHFFFAOYSA-N
Synonym 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
PubChem CID 3084924
IUPAC Name 1-benzylpiperidin-3-one;hydrochloride
SMILES Cl.O=C1CCCN(CC2=CC=CC=C2)C1

Specifications

Melting Point 105°C to 107°C
Quantity 25 g
Beilstein 3705146
Formula Weight 225.72 (Anhydrous)
Chemical Name or Material 1-Benzyl-3-piperidone hydrochloride hydrate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 256-646-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.