Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-hydroxypiperidine 98.0+%, TCI America™
SDP

Catalog No. B22525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22525G 5 g
B225225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22525G Supplier TCI America Supplier No. B22525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4727-72-4
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00006503
InChI Key BPPZXJZYCOETDA-UHFFFAOYSA-N
Synonym 1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
PubChem CID 78461
IUPAC Name 1-benzylpiperidin-4-ol
SMILES C1CN(CCC1O)CC2=CC=CC=C2

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 191.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.