Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | 189.25 | 1 ML
SDP

Catalog No. 5000448616
Encompass_Preferred

1-Benzyl-4-piperidone is a laboratory chemical with a molecular weight of 189.25, identified by CAS number 3612-20-2. It is primarily used for research purposes and in the manufacture of substances.

  • Used as a laboratory chemical
  • Used in the manufacture of substances
  • Formula: C12H15NO
  • Appearance: Liquid
  • Boiling point/range: 278.5°C at 760 mmHg
  • Flash point: 71°C
  • Relative density: 1.021 g/cm3
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Recommended storage temperature (pure form): -20°C for 3 years, 4°C for 2 years
  • Recommended storage temperature (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks

Catalog No. 50-004-48616 Supplier Medchemexpress LLC Supplier No. HYW00782210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Benzyl-4-piperidone is a laboratory chemical with a molecular weight of 189.25, identified by CAS number 3612-20-2. It is primarily used for research purposes and in the manufacture of substances.

  • Used as a laboratory chemical
  • Used in the manufacture of substances
  • Formula: C12H15NO
  • Appearance: Liquid
  • Boiling point/range: 278.5°C at 760 mmHg
  • Flash point: 71°C
  • Relative density: 1.021 g/cm3
  • Keep container tightly sealed in cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Recommended storage temperature (pure form): -20°C for 3 years, 4°C for 2 years
  • Recommended storage temperature (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.