Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | 189.25 | 100 G
SDP

Catalog No. 5000448598
Encompass_Preferred

1-Benzyl-4-piperidone is a laboratory chemical primarily used for the manufacture of various substances. This compound has an appearance of a liquid, a molecular formula of C12H15NO, and a molecular weight of 189.25. It is intended for research use only by experienced personnel.

  • Identified for use as a laboratory chemical
  • Used in the manufacture of substances
  • Liquid appearance
  • Boiling point of 278.5°C at 760 mmHg
  • Flash point of 71°C
  • Relative density of 1.021 g/cm³

Catalog No. 50-004-48598 Supplier Medchemexpress LLC Supplier No. HYW007822100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Benzyl-4-piperidone is a laboratory chemical primarily used for the manufacture of various substances. This compound has an appearance of a liquid, a molecular formula of C12H15NO, and a molecular weight of 189.25. It is intended for research use only by experienced personnel.

  • Identified for use as a laboratory chemical
  • Used in the manufacture of substances
  • Liquid appearance
  • Boiling point of 278.5°C at 760 mmHg
  • Flash point of 71°C
  • Relative density of 1.021 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.