Learn More
Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | MFCD00006192 | 189.25 | 25 G
Supplier: Medchemexpress LLC 25GHYW007822
1-Benzyl-4-piperidone serves as a versatile heterocyclic building block in the synthesis of pharmaceutical compounds.
- Used as a building block in the synthesis of various medicinal compounds.
- Synthesizes spirocyclic furopyridines as haloperidol-sensitive σ receptor ligands.
- Synthesizes multi-target-directed donepezil + propargylamine + 8-hydroxyquinoline (DPH) hybrids for Alzheimer's disease treatment.
- Synthesizes spiropiperidine iminohydantoin aspartyl protease inhibitors.