Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Benzyl-4-piperidone | 3612-20-2 | MFCD00006192 | 189.25 | 500 G
SDP

Catalog No. 5000447614
Encompass_Preferred

1-Benzyl-4-piperidone is a chemical substance primarily used in laboratory research and for the manufacture of other substances. This compound is a combustible liquid and is classified as harmful if swallowed, and can cause skin and serious eye irritation. It has a molecular formula of C12H15NO.

  • Appears as a liquid
  • Molecular weight is 189.25
  • Has a minimum purity of >95%
  • Boiling point is 278.5°C at 760 mmHg
  • Flash point is 71°C
  • Store tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition
  • For pure form, store at -20°C for up to 3 years, or 4°C for up to 2 years
  • When in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month

Catalog No. 50-004-47614 Supplier Medchemexpress LLC Supplier No. HYW007822500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Benzyl-4-piperidone is a chemical substance primarily used in laboratory research and for the manufacture of other substances. This compound is a combustible liquid and is classified as harmful if swallowed, and can cause skin and serious eye irritation. It has a molecular formula of C12H15NO.

  • Appears as a liquid
  • Molecular weight is 189.25
  • Has a minimum purity of >95%
  • Boiling point is 278.5°C at 760 mmHg
  • Flash point is 71°C
  • Store tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition
  • For pure form, store at -20°C for up to 3 years, or 4°C for up to 2 years
  • When in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.