Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Benzyl-4-piperidyl)methanol, Thermo Scientific™, Thermo Scientific™

Catalog No. KM09623DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM09623DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM09623DE Supplier Thermo Scientific Chemicals Supplier No. KM09623DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67686-01-5
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD00831014
InChI Key FLQPYEOKVZYXRL-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidyl methanol, 1-benzylpiperidin-4-yl methanol, 1-benzyl-4-piperidinemethanol, 1-benzyl-4-piperidinyl methanol, 1-benzyl-4-hydroxymethylpiperidine, 4-piperidinemethanol, 1-phenylmethyl, 1-benzy-4-hydroxymethylpiperidine, 1-benzy-4-hydroxymethyl piperidine, pubchem8894, 1-benzyl-4-piperdinemethanol
PubChem CID 736802
IUPAC Name (1-benzylpiperidin-4-yl)methanol
SMILES C1CN(CCC1CO)CC2=CC=CC=C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 205.3
Percent Purity 97%
Chemical Name or Material (1-Benzyl-4-piperidyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.