Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl D-Aspartate 98.0+%, TCI America™
SDP

Catalog No. B29931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29931G 1 g
B29935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29931G Supplier TCI America Supplier No. B29931G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79337-40-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
InChI Key NJSRYBIBUXBNSW-SECBINFHSA-N
Synonym 1-benzyl d-aspartate, d-aspartic acid alpha-benzyl ester, 3r-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3r-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyld-aspartate, d-aspartic acid, a-benzyl ester, d-aspartic acid 1-benzyl ester, d-aspartic acid, 1-phenylmethyl ester
PubChem CID 6995008
IUPAC Name (3R)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N

Specifications

Melting Point 176°C
Color White
Quantity 1 g
Formula Weight 223.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl D-Aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.