Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™
SDP

Catalog No. B24085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24085G 5 g
B240825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24085G Supplier TCI America Supplier No. B24085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30925-18-9
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
InChI Key LDRWTKQWSXGSTM-LBPRGKRZSA-N
Synonym boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn
PubChem CID 7017887
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 99°C
Color White
Quantity 5 g
Formula Weight 323.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.