Learn More
1-Benzyloxy-2-iodoethane, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Linear Formula | C6H5CH2OCH2CH2I |
| Quantity | 250 mg |
| UN Number | UN2810 |
| IUPAC Name | [(2-iodoethoxy)methyl]benzene |
| Formula Weight | 262.09 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Benzyloxy-2-iodoethane |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H301-H311-H331 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. |
| Precautionary Statement | GHS P Statement P261-P280-P301+P310-P361-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Remove/Take off immediately all contaminated clothing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.