Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyloxycarbonyl-3-piperidinemethanol, 97%, Thermo Scientific™

Catalog No. AAH3528503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3528503 1 g
AAH3528506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3528503 Supplier Thermo Scientific Chemicals Supplier No. H3528503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39945-51-2
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD03407298
InChI Key XLWOOUZKMJBINO-UHFFFAOYSA-N
Synonym benzyl 3-hydroxymethyl piperidine-1-carboxylate, 1-cbz-3-hydroxymethylpiperidine, n-cbz-3-piperidinemethanol, n-cbz-3-hydroxymethylpiperidine, benzyl 3-hydroxymethyl tetrahydro-1 2h-pyridinecarboxylate, 3-hydroxymethyl-piperidine-1-carboxylic acid benzyl ester, 1-piperidinecarboxylic acid, 3-hydroxymethyl-, phenylmethyl ester, 1-benzyloxycarbonyl-3-piperidinemethanol, pubchem11832, 1-n-cbz-3-piperidinemethanol
PubChem CID 2776577
IUPAC Name benzyl 3-(hydroxymethyl)piperidine-1-carboxylate
SMILES C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)CO

Specifications

Melting Point 59°C to 60°C
Quantity 1 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material 1-Benzyloxycarbonyl-3-piperidinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.