Learn More
Medchemexpress LLC 1-beta-D-Arabinofuranosyluracil | 3083-77-0 | 10 MM * 1 ML
SDP
Catalog No. 5000418450
Encompass_Preferred
1-beta-D-Arabinofuranosyluracil is a methoxyadenosine derivative isolated from the Caribbean sponge *Tectitethya crypta*. It demonstrates a diverse bioactivity profile, including anti-inflammatory, analgesic, and vasodilation properties. It also reduces the proliferation of mouse lymphoma cells.
  • Exhibits anti-inflammatory activity
  • Possesses analgesic and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Shows cytostatic activity against various human cell lines
  • Strongly inhibits RNA-directed DNA polymerases
Catalog No. 50-004-18450 Supplier Medchemexpress LLC Supplier No. HYN665210MM/1ML
1-beta-D-Arabinofuranosyluracil is a methoxyadenosine derivative isolated from the Caribbean sponge *Tectitethya crypta*. It demonstrates a diverse bioactivity profile, including anti-inflammatory, analgesic, and vasodilation properties. It also reduces the proliferation of mouse lymphoma cells.
  • Exhibits anti-inflammatory activity
  • Possesses analgesic and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Shows cytostatic activity against various human cell lines
  • Strongly inhibits RNA-directed DNA polymerases