Learn More
Medchemexpress LLC 1-beta-D-Arabinofuranosyluracil | 3083-77-0 | 50 MG
SDP
Catalog No. 5000420178
Encompass_Preferred
1-beta-D-Arabinofuranosyluracil (Standard) is an analytical standard derived from the Caribbean sponge Tectitethya crypta. This methoxyadenosine derivative is intended for research and analytical applications, offering a diverse bioactivity profile including anti-inflammatory, analgesic, and vasodilation properties. It has also been shown to reduce the proliferation of mouse lymphoma cells. This compound is a reference standard commonly used in qualitative, quantitative, and methodological research experiments across techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Isolated from the Caribbean sponge Tectitethya crypta
  • Exhibits anti-inflammatory activity
  • Possesses analgesic and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Used as an analytical standard in HPLC, GC, and MS
Catalog No. 50-004-20178 Supplier Medchemexpress LLC Supplier No. HYN6652R50MG
1-beta-D-Arabinofuranosyluracil (Standard) is an analytical standard derived from the Caribbean sponge Tectitethya crypta. This methoxyadenosine derivative is intended for research and analytical applications, offering a diverse bioactivity profile including anti-inflammatory, analgesic, and vasodilation properties. It has also been shown to reduce the proliferation of mouse lymphoma cells. This compound is a reference standard commonly used in qualitative, quantitative, and methodological research experiments across techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Isolated from the Caribbean sponge Tectitethya crypta
  • Exhibits anti-inflammatory activity
  • Possesses analgesic and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Used as an analytical standard in HPLC, GC, and MS