Learn More
Medchemexpress LLC 1-beta-D-Arabinofuranosyluracil (Standard) | 3083-77-0 | 25 MG
SDP
Catalog No. 5000421626
Encompass_Preferred
1-beta-D-Arabinofuranosyluracil (Standard) is an analytical standard for research and analytical applications, isolated from the Caribbean sponge *Tectitethya crypta*. This methoxyadenosine derivative exhibits diverse bioactivity, including anti-inflammatory, analgesic, and vasodilation properties. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Isolated from Caribbean sponge *Tectitethya crypta*
  • Exhibits anti-inflammatory, analgesic, and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Used in HPLC, GC, and MS research experiments
  • Store at 4°C, sealed, away from light and moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from light and moisture)
Catalog No. 50-004-21626 Supplier Medchemexpress LLC Supplier No. 25MGHYN6652R
1-beta-D-Arabinofuranosyluracil (Standard) is an analytical standard for research and analytical applications, isolated from the Caribbean sponge *Tectitethya crypta*. This methoxyadenosine derivative exhibits diverse bioactivity, including anti-inflammatory, analgesic, and vasodilation properties. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Isolated from Caribbean sponge *Tectitethya crypta*
  • Exhibits anti-inflammatory, analgesic, and vasodilation properties
  • Reduces proliferation of mouse lymphoma cells
  • Used in HPLC, GC, and MS research experiments
  • Store at 4°C, sealed, away from light and moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from light and moisture)