Learn More
AdipoGen DL-3-Hydroxyisobutyric acid sodium salt
SDP
Supplier:  AdipoGen CDXH0085M250
Encompass_Preferred
DL-3-Hydroxyisobutyric acid sodium salt, AdipoGen Life Sciences. Chemical. CAS: 1219589-99-7. Formula: C4H7NaO3. MW: 126.09. Synthetic Intermediate formed in the valine catabolism. It is a reactant of the enzymes involved in branched chain amino acid metabolism 3-hydroxyisobutyrate dehydrogenase and 3-hydroxyisobutyryl-CoA hydrolase and it was found to have increased concentrations in MS patient metabolic profiles. This compound may be used to study the distribution, characterization and kinetics of these enzymes. Shown to inhibit key enzymes of the energy metabolism.
Catalog No. 50-114-7132