Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[Bis(2-hydroxyethyl)amino]-2-propanol 93.0+%, TCI America™
SDP

Catalog No. B21805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B21805G Supplier TCI America Supplier No. B21805G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 6712-98-7
Molecular Formula C7H17NO3
Molecular Weight (g/mol) 163.22
MDL Number MFCD00009725
InChI Key ZFECCYLNALETDE-UHFFFAOYNA-N
Synonym 1-bis 2-hydroxyethyl amino-2-propanol, 1-bis 2-hydroxyethyl amino propan-2-ol, 2-propanol, 1-bis 2-hydroxyethyl amino, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, n,n-bis 2-hydroxyethyl isopropanolamine, 2,2'-2-hydroxypropyl azanediyl diethanol, acmc-209nxu, 1-n,n-bis 2-hydroxyethyl amino propan-2-ol
PubChem CID 97712
IUPAC Name 1-[bis(2-hydroxyethyl)amino]propan-2-ol
SMILES CC(O)CN(CCO)CCO

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 163.22
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-[Bis(2-hydroxyethyl)amino]-2-propanol
EINECSNumber (2)-0353
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.