Learn More
Medchemexpress LLC 1-[bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide hexafluorophosphate | 148893-10-1 | MFCD00274639 | 100g
SDP
Catalog No. 5000266143
Encompass_Preferred
HATU (1-[Bis(dimethylamino)methylene]-1H-1 2 3-triazolo[4 5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used in peptide coupling chemistry to generate an active ester from a carboxylic acid HATU can be used along with H nig s base (N N-diisopropylethylamine DIPEA) to form amide bonds[1]
Catalog No. 50-002-66143 Supplier Medchemexpress LLC Supplier No. HYY1703100G
HATU (1-[Bis(dimethylamino)methylene]-1H-1 2 3-triazolo[4 5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used in peptide coupling chemistry to generate an active ester from a carboxylic acid HATU can be used along with H nig s base (N N-diisopropylethylamine DIPEA) to form amide bonds[1]