Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[Bis(trifluoromethanesulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene 98.0+%, TCI America™
SDP

Catalog No. B2291100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2291100MG 100 mg
B22911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2291100MG Supplier TCI America Supplier No. B2291100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 405074-81-9
Molecular Formula C9HF11O4S2
Molecular Weight (g/mol) 446.205
MDL Number MFCD04117910
InChI Key RLLDXJXYMKTGPV-UHFFFAOYSA-N
Synonym alpha,alpha-Bis(trifluoromethanesulfonyl)-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorophenylbis(trifluoromethanesulfonyl)methane
PubChem CID 10203986
IUPAC Name 1-[bis(trifluoromethylsulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 89°C
Quantity 100 mg
UN Number 3261
Formula Weight 446.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[Bis(trifluoromethanesulfonyl)methyl]-2,3,4,5,6-pentafluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.