Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-2-piperidone | 85908-96-9 | >=98.0% | 199.25 | 10 G
SDP

Catalog No. 5000404342
Encompass_Preferred

1-Boc-2-piperidone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a solid, ranging from white to light brown.

  • Molecular weight of 199.25
  • Purity of 98.0%
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Solubility in DMSO is 200 mg/mL

Catalog No. 50-004-04342 Supplier Medchemexpress LLC Supplier No. HY4002110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Boc-2-piperidone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It appears as a solid, ranging from white to light brown.

  • Molecular weight of 199.25
  • Purity of 98.0%
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Solubility in DMSO is 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.