Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-1-Boc-3-Aminopyrrolidine

Catalog No. 6440645G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-406-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 64-406-45G Supplier Sigma Aldrich Supplier No. 6440645G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-1-Boc-3-aminopyrrolidine may be used to prepare:Descarboxamide analog and Na-methylated analogs of 4-N-(N-d-nitro-L-argininyl)-trans-4-amino-L-proline amide.Histamine 3 (H3) receptor antagonists containing pyrrolidin-3-yl-N-methylbenzamide moiety.

Specifications

CAS 871125-86-9
Melting Point 82°C to 86°C
Molecular Formula C11H19NO5
Refractive Index n20/D 1.472
Linear Formula C11H19NO5
MDL Number MFCD00237541
Quantity 5 g
Synonym (R)-(+)-N-Boc-3-aminopyrrolidine
Molecular Weight (g/mol) 245.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.