Learn More
Accela Chembio Inc 1-boc-3-azetidinone | 5g | 398489-26-4 | MFCD01861741 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon
Supplier:  Accela Chembio Inc SY0040165G
Encompass_Preferred
1-boc-3-azetidinone | 5g | 398489-26-4 | MFCD01861741 | 97+% | Shelf Life: 1620 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon
Catalog No. 50-250-346