Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-1-Boc-3-bromopyrrolidine, 95%

Catalog No. AAH5707103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5707103 1 g
AAH5707106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5707103 Supplier Thermo Scientific Chemicals Supplier No. H5707103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 939793-16-5 | C9H16BrNO2 | 250.14 g/mol

(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939793-16-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD08752478
InChI Key QJTKPXFJOXKUEY-UHFFFAOYNA-N
PubChem CID 42614217
IUPAC Name tert-butyl 3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Br)C1

Specifications

Melting Point 45°C to 53°C
Quantity 1 g
UN Number UN3077
Synonym n-boc-3-bromopyrrolidine, 1-boc-3-bromopyrrolidine, 3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, +/--1-boc-3-bromopyrrolidine, acmc-209zg9, ^+-1-boc-3-bromopyrrolidine, rs 1-tertbutoxycarbonyl-3-bromopyrrolidine, rs 1-tert-butoxycarbonyl-3-bromopyrrolidine, 2-methyl-2-propanyl 3-bromo-1-pyrrolidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (+/-)-1-Boc-3-bromopyrrolidine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.