Learn More
Medchemexpress LLC 1-Boc-3-(hydroxymethyl)pyrrolidine | 114214-69-6 | 98.5% | C10H19NO3 | 1 ML
SDP
Catalog No. 5000434716
Encompass_Preferred
1-Boc-3-(hydroxymethyl)pyrrolidine is an intermediate used to synthesize a FGFR1/2/3/4 inhibitor (Compound 2). It is also utilized in cancer research.
  • Used to synthesize a FGFR1/2/3/4 inhibitor
  • Utilized in cancer research
Catalog No. 50-004-34716 Supplier Medchemexpress LLC Supplier No. HY6021810MM/1ML
1-Boc-3-(hydroxymethyl)pyrrolidine is an intermediate used to synthesize a FGFR1/2/3/4 inhibitor (Compound 2). It is also utilized in cancer research.
  • Used to synthesize a FGFR1/2/3/4 inhibitor
  • Utilized in cancer research