Learn More
Medchemexpress LLC 1-Boc-3-(hydroxymethyl)pyrrolidine | 114214-69-6 | 98.5% | C10H19NO3 | 10 G
SDP
Catalog No. 5000434740
Encompass_Preferred
1-Boc-3-(hydroxymethyl)pyrrolidine is an intermediate used in cancer research. It serves as a precursor for synthesizing a FGFR1/2/3/4 inhibitor (Compound 2).
  • Intermediate for FGFR1/2/3/4 inhibitor synthesis
  • Applicable in cancer research
  • Appears as a light yellow to light brown liquid
  • Density of 1.089 g/cm³
Catalog No. 50-004-34740 Supplier Medchemexpress LLC Supplier No. HY6021810G
1-Boc-3-(hydroxymethyl)pyrrolidine is an intermediate used in cancer research. It serves as a precursor for synthesizing a FGFR1/2/3/4 inhibitor (Compound 2).
  • Intermediate for FGFR1/2/3/4 inhibitor synthesis
  • Applicable in cancer research
  • Appears as a light yellow to light brown liquid
  • Density of 1.089 g/cm³