Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-3-methylazetidine, 95%, Thermo Scientific Chemicals

Catalog No. AAH57496ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57496ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57496ME Supplier Thermo Scientific Chemicals Supplier No. H57496ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1314984-00-3 | C10H15NO2 | 181.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314984-00-3
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD17016080
InChI Key UENGYBYGCXKNRF-UHFFFAOYSA-N
Synonym 1-boc-3-ethynylazetidine, 3-ethynyl-1-azetidinecarboxylic acid tert-butyl ester, 1-boc-3-ethynyl-azetidine, tert-butyl3-ethynylazetidine-1-carboxylate, 1-azetidinecarboxylic acid, 3-ethynyl-, 1,1-dimethylethyl ester
PubChem CID 54349237
IUPAC Name tert-butyl 3-ethynylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C#C

Specifications

Quantity 500 mg
Beilstein 15408998
Formula Weight 171.24
Percent Purity 95%
Chemical Name or Material 1-Boc-3-methylazetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.