Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-3-piperidone, 97%

Catalog No. AAL1927603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1927603 1 g
AAL1927606 5 g
AAL1927614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1927603 Supplier Thermo Scientific Chemicals Supplier No. L1927603
Add to Cart

CAS: 98977-36-7 | C10H17NO3 | 199.25 g/mol

1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98977-36-7
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01631193
InChI Key RIFXIGDBUBXKEI-UHFFFAOYSA-N
Synonym 1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
PubChem CID 2756825
IUPAC Name tert-butyl 3-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)C1

Specifications

Melting Point 37°C to 39°C
Quantity 1 g
Beilstein 5936353
Solubility Information It is insoluble in water.
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 1-Boc-3-piperidone
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.