Learn More
Medchemexpress LLC 1-Boc-3-piperidone | 98977-36-7 | MFCD01631193 | >98.5% | 199.25 | 25 G
SDP
Catalog No. 5000403028
Encompass_Preferred
1-Boc-3-piperidone is a Boc-protected piperidone derivative and an important intermediate in organic synthesis. It can be used to study compounds related to inflammatory skin diseases (such as atopic dermatitis), lymphoma, and other diseases.
  • Boc-protected piperidone derivative.
  • Important intermediate in organic synthesis.
  • Can be used to study compounds related to inflammatory skin diseases (e.g., atopic dermatitis).
  • Applicable in research concerning lymphoma and other diseases.
Catalog No. 50-004-03028 Supplier Medchemexpress LLC Supplier No. 25GHY40020
1-Boc-3-piperidone is a Boc-protected piperidone derivative and an important intermediate in organic synthesis. It can be used to study compounds related to inflammatory skin diseases (such as atopic dermatitis), lymphoma, and other diseases.
  • Boc-protected piperidone derivative.
  • Important intermediate in organic synthesis.
  • Can be used to study compounds related to inflammatory skin diseases (e.g., atopic dermatitis).
  • Applicable in research concerning lymphoma and other diseases.