Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-4-(2-hydroxyethyl)piperazine-d4 | 1219802-10-4 | 99.0% | 234.33 | C11H18D4N2O3 | 10 MG
SDP

Catalog No. 5000297760
Encompass_Preferred

1-BOC-4-(2-hydroxyethyl)piperazine-d4 is the deuterium-labeled analogue of 1-BOC-4-(2-hydroxyethyl)piperazine, provided as a stable isotope reagent for laboratory research. It is intended for use as an internal standard or tracer in analytical chemistry and mass spectrometry workflows. For research use only; not for human or clinical use.

  • Deuterium-labeled analogue for use as an internal standard in mass spectrometry.
  • High purity confirmed by HPLC (99.0%).
  • Molecular formula C11H18D4N2O3 and molecular weight 234.33.
  • Available in small quantities suitable for analytical applications.
  • Stable solid form with recommended storage at -20 °C for long-term stability.
  • Supplied with a certificate of analysis for batch-specific confirmation.

Catalog No. 50-002-97760 Supplier Medchemexpress LLC Supplier No. HY20797S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-BOC-4-(2-hydroxyethyl)piperazine-d4 is the deuterium-labeled analogue of 1-BOC-4-(2-hydroxyethyl)piperazine, provided as a stable isotope reagent for laboratory research. It is intended for use as an internal standard or tracer in analytical chemistry and mass spectrometry workflows. For research use only; not for human or clinical use.

  • Deuterium-labeled analogue for use as an internal standard in mass spectrometry.
  • High purity confirmed by HPLC (99.0%).
  • Molecular formula C11H18D4N2O3 and molecular weight 234.33.
  • Available in small quantities suitable for analytical applications.
  • Stable solid form with recommended storage at -20 °C for long-term stability.
  • Supplied with a certificate of analysis for batch-specific confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.